3-tert-Butylphenol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-tert-Butylphenol

B24315-06 B24315-14 B24315-22
CAAAB24315-06EA 104.44 CAD
CAAAB24315-06 CAAAB24315-14 CAAAB24315-22
3-tert-Butylphenol 99%
3-tert-Butylphenol

Purity: 99% CAS No.: 585-34-2 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 108°(228°F) Melting Point: 44-46° Boiling Point: 125-130°/20mm UN No.: 2430 MDL No.: MFCD00002300

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 125…130 °C/20 mm
Melting Pt: 44…46 °C
Density: 1.048 g/cm³ (20 °C)
Flash Pt: 108 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002300
CAS Number: 585-34-2
EINECS: 209-553-4
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR