2,3,5,6-Tetrafluoroanisole 97%

Supplier: Thermo Scientific Chemicals
Warning

B24275-06 B24275-14
CAAAB24275-06EA 81.69 CAD
CAAAB24275-06 CAAAB24275-14 CAAAB24275-22
2,3,5,6-Tetrafluoroanisole 97%
2,3,5,6-Tetrafluoroanisole

Purity: 97% CAS No.: 2324-98-3 Molecular Formula: C7H4F4O Formula Weight: 180.10 Flash Point: 41°(107°F) Refractive Index: 1.4280 Boiling Point: 144-145° UN No.: 1993 MDL No.: MFCD00000311

Formula: C₇H₄F₄O
Boiling Pt: 144…145 °C
Density: 1.293
Flash Pt: 41 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000311
CAS Number: 2324-98-3
EINECS: 219-037-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR