3-Ethoxyphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

B24266-06 B24266-14 B24266-22
CAAAB24266-06EA 102.38 CAD
CAAAB24266-06 CAAAB24266-14 CAAAB24266-22
3-Ethoxyphenol 98%
3-Ethoxyphenol

Purity: 98% CAS No.: 621-34-1 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 93°(199°F) Refractive Index: 1.5410 Odor: Phenol-like Melting Point: 28-31° Boiling Point: 247° MDL No.: MFCD00016450

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 247 °C
Melting Pt: 28…31 °C
Density: 1.070
Flash Pt: >93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016450
CAS Number: 621-34-1
EINECS: 210-681-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR