(±)-4-Chloromandelic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloromandelic acid, (±)-4-Chlorophenylglycolic acid

B24264-06 B24264-14
CAAAB24264-06EA 83.27 CAD
CAAAB24264-06 CAAAB24264-14
(±)-4-Chloromandelic acid 98%
(±)-4-Chloromandelic acid

Purity: 98% CAS No.: 492-86-4 Molecular Formula: C8H7ClO3 Formula Weight: 186.60 Melting Point: 117-122° Beilstein Reference: 1567759

Formula: C₈H₇ClO₃
MW: 186.59 g/mol
Boiling Pt: 361
Melting Pt: 117…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00042724
CAS Number: 492-86-4
EINECS: 207-764-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR