2-Hydroxyethyl methacrylate 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) ~ 200 ppm
B24260-22
B24260-36
CAAAB24260-22EA
42.19
CAD
CAAAB24260-22
CAAAB24260-36
2-Hydroxyethyl methacrylate 97% stabilized
2-Hydroxyethyl methacrylate
Purity: 97%
CAS No.: 868-77-9
Molecular Formula: CH2=CH(CH3)CO2CH2CH2OH
Formula Weight: 130.14
Storage Temperature: 2°C to 8°C
Flash Point: 101°(213°F)
Refractive Index: 1.4530
Odor: Ester-like
Melting Point: -12°
Boiling Point: 250°
Beilstein Reference: 1071583
MDL No.: MFCD00002863
Formula:
C₆H₁₀O₃ Boiling Pt: 250 °C Melting Pt: –12 °C Density: 1.071 Flash Pt: 101 °C(213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002863 CAS Number: 868-77-9 EINECS: 212-782-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...