2,6-Dimethoxyphenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24253-14
B24253-22
CAAAB24253-14EA
77.89
CAD
CAAAB24253-14
CAAAB24253-22
2,6-Dimethoxyphenol 99%
2,6-Dimethoxyphenol
Purity: 99%
CAS No.: 91-10-1
Molecular Formula: C8H10O3
Formula Weight: 154.16
Flash Point: 140°(284°F)
Melting Point: 50-57°
Boiling Point: 261°
Beilstein Reference: 1526871
MDL No.: MFCD00064434
Formula:
C₈H₁₀O₃ MW: 154.17 g/mol Boiling Pt: 261 °C Melting Pt: 50…57 °C Flash Pt: 140 °C(284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064434 CAS Number: 91-10-1 EINECS: 202-041-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...