3,4-Dimethoxyphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24240-03
B24240-06
B24240-14
CAAAB24240-03EA
58.78
CAD
CAAAB24240-03
CAAAB24240-06
CAAAB24240-14
3,4-Dimethoxyphenylboronic acid 98%
3,4-Dimethoxyphenylboronic acid
Purity: 98%
CAS No.: 122775-35-3
Molecular Formula: C8H11BO4
Formula Weight: 181.98
Melting Point: ca 240°
Beilstein Reference: 5334877
Formula:
C₈H₁₁BO₄ MW: 181.98 Melting Pt: ∼ 240 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074574 CAS Number: 122775-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...