4-Nitro-o-anisidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24220-14
B24220-22
B24220-36
CAAAB24220-14EA
36.66
CAD
CAAAB24220-14
CAAAB24220-22
CAAAB24220-36
4-Nitro-o-anisidine 99%
4-Nitro-o-anisidine
Purity: 99%
CAS No.: 97-52-9
Molecular Formula: C7H8N2O3
Formula Weight: 168.15
Melting Point: 138-142°
Beilstein Reference: 879619
Formula:
C₇H₈N₂O₃ Boiling Pt: 364 °C (1013 hPa) Melting Pt: 138…142 °C Density: 1.211 Storage Temperature: Ambient |
MDL Number:
MFCD00007363 CAS Number: 97-52-9 EINECS: 202-588-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...