1-Iodoheptane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with copper

B24206-18 B24206-30
CAAAB24206-18EA 57.35 CAD
CAAAB24206-18 CAAAB24206-30
1-Iodoheptane 98% stabilized
1-Iodoheptane

Purity: 98% CAS No.: 4282-40-0 Molecular Formula: CH3(CH2)6I Formula Weight: 226.10 Flash Point: 78°(174°F) Refractive Index: 1.4900 Melting Point: -48° Boiling Point: 203-205° MDL No.: MFCD00001104

Formula: C₇H₁₅I
Boiling Pt: 203…205 °C
Melting Pt: –48 °C
Density: 1.379
Flash Pt: 78 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001104
CAS Number: 4282-40-0
EINECS: 224-285-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR