Ethoxyacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24194-14
B24194-22
CAAAB24194-14EA
126.56
CAD
CAAAB24194-14
CAAAB24194-22
Ethoxyacetic acid 98%
Ethoxyacetic acid
Purity: 98%
CAS No.: 627-03-2
Molecular Formula: CH3CH2OCH2CO2H
Formula Weight: 104.11
Flash Point: 97°(206°F)
Refractive Index: 1.4190
Odor: Pungent
Boiling Point: 207°
Beilstein Reference: 1743037
UN No.: 3265
MDL No.: MFCD00004310
Formula:
C₄H₈O₃ Boiling Pt: 207 °C Density: 1.102 Flash Pt: 97 °C(206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004310 CAS Number: 627-03-2 EINECS: 210-978-2 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...