6-Methyluracil 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Methylpyrimidine-2,4(1H,3H)-dione, 4-Methyluracil, 6-Methyl-2,4(1H,3H)-pyrimidinedione
B24191-0E
B24191-22
B24191-36
CAAAB24191-0EEA
625.68
CAD
CAAAB24191-0E
CAAAB24191-22
CAAAB24191-36
6-Methyluracil 97%
6-Methyluracil
Purity: 97%
CAS No.: 626-48-2
Molecular Formula: C5H6N2O2
Formula Weight: 126.12
Melting Point: 317-320°
Beilstein Reference: 115647
Merck Reference: 14,6133
MDL No.: MFCD00006028
Formula:
C₅H₆N₂O₂ MW: 126.11 g/mol Boiling Pt: 358 °C (1013 hPa) Melting Pt: 317…320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006028 CAS Number: 626-48-2 EINECS: 210-949-4 Merck Index: 14,6133 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...