N-Butylaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

B24180-14 B24180-22
CAAAB24180-14EA 109.65 CAD
CAAAB24180-14 CAAAB24180-22
N-Butylaniline 99%
N-Butylaniline

Purity: 99% CAS No.: 1126-78-9 Molecular Formula: C10H15N Formula Weight: 149.23 Flash Point: 105°(222°F) Refractive Index: 1.5320 Boiling Point: 240-242° UN No.: 2738 MDL No.: MFCD00035798

Formula: (C₆H₅)NHCH₂CH₂CH₂CH₃
MW: 149.24 g/mol
Boiling Pt: 240…242 °C
Melting Pt: –14.4 °C
Density: 0.932
Flash Pt: 105 °C (222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00035798
CAS Number: 1126-78-9
EINECS: 214-425-6
UN: 2738
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR