Ethyl iodoacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24177-06
B24177-14
CAAAB24177-06EA
111.86
CAD
CAAAB24177-06
CAAAB24177-14
Ethyl iodoacetate 98%
Ethyl iodoacetate
Purity: 98%
CAS No.: 623-48-3
Molecular Formula: ICH2CO2CH2CH3
Formula Weight: 214
Storage Temperature: 2°C to 8°C
Flash Point: 76°(169°F)
Refractive Index: 1.503
Boiling Point: 179-180°
Beilstein Reference: 741934
UN No.: 2927
MDL No.: MFCD00001081
Formula:
C₄H₇IO₂ MW: 214 g/mol Boiling Pt: 179 Density: 1.81 Flash Pt: 76 °C (169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001081 CAS Number: 623-48-3 EINECS: 210-796-3 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...