2-Naphthyl boronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24157-03
B24157-06
B24157-14
CAAAB24157-03EA
45.98
CAD
CAAAB24157-03
CAAAB24157-06
CAAAB24157-14
2-Naphthyl boronic acid 97%
2-Naphthyl boronic acid
Purity: 97%
CAS No.: 32316-92-0
Molecular Formula: C10H9BO2
Formula Weight: 171.99
Storage Temperature: 2°C to 8°C
Melting Point: ca 267°
Beilstein Reference: 2936449
MDL No.: MFCD00236051
Formula:
C₁₀H₉BO₂ Melting Pt: ∼ 267 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236051 CAS Number: 32316-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...