3-(Trifluoromethyl)benzylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24143-06
B24143-14
CAAAB24143-06EA
96.06
CAD
CAAAB24143-06
CAAAB24143-14
3-(Trifluoromethyl)benzylamine 97%
3-(Trifluoromethyl)benzylamine
Purity: 97%
CAS No.: 2740-83-2
Molecular Formula: C8H8F3N
Formula Weight: 175.15
Flash Point: 71°(159°F)
Refractive Index: 1.4630
Boiling Point: 93-97°/28mm
Beilstein Reference: 1102465
UN No.: 2735
MDL No.: MFCD00008117
Formula:
C₈H₈F₃N Boiling Pt: 93…97 °C/28 mm Density: 1.222 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008117 CAS Number: 2740-83-2 EINECS: 220-367-2 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...