4-Methylimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methyl-1H-imidazole

B24139-09 B24139-18 B24139-30
CAAAB24139-09EA 45.66 CAD
CAAAB24139-09 CAAAB24139-18 CAAAB24139-30
4-Methylimidazole 98%
4-Methylimidazole

Purity: 98% CAS No.: 822-36-6 Molecular Formula: C4H6N2 Formula Weight: 82.11 Flash Point: 157°(314°F) Melting Point: 49-57° Boiling Point: 263° Beilstein Reference: 1453 UN No.: 3263 MDL No.: MFCD00005201

Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 263 °C
Melting Pt: 49…57 °C
Density: 1.020
Flash Pt: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005201
CAS Number: 822-36-6
EINECS: 212-497-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR