tert-Butylcyclohexane 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24126-14
B24126-22
CAAAB24126-14EA
84.85
CAD
CAAAB24126-14
CAAAB24126-22
tert-Butylcyclohexane 99+%
tert-Butylcyclohexane
Purity: 99+%
CAS No.: 3178-22-1
Molecular Formula: C10H20
Formula Weight: 140.27
Flash Point: 40°(104°F)
Refractive Index: 1.4470
Melting Point: -41°
Boiling Point: 171°
UN No.: 3295
MDL No.: MFCD00001471
Formula:
C₁₀H₂₀ Boiling Pt: 171 °C Melting Pt: –41 °C Density: 0.830 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001471 CAS Number: 3178-22-1 EINECS: 221-652-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...