2,3-Dimethoxyphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B24125-06 B24125-14
CAAAB24125-03EA 76.63 CAD
CAAAB24125-03 CAAAB24125-06 CAAAB24125-14
2,3-Dimethoxyphenylboronic acid 98%
2,3-Dimethoxyphenylboronic acid

Purity: 98% CAS No.: 40972-86-9 Molecular Formula: C8H11BO4 Formula Weight: 181.98 Melting Point: 67-71° Beilstein Reference: 3051295

Formula: C₈H₁₁BO₄
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD02683112
CAS Number: 40972-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR