1,3-Dibromobutane 98%

Supplier: Thermo Scientific Chemicals
Warning

B24123-14 B24123-22
CAAAB24123-14EA 60.2 CAD
CAAAB24123-14 CAAAB24123-22
1,3-Dibromobutane 98%
1,3-Dibromobutane

Purity: 98% CAS No.: 107-80-2 Molecular Formula: C4H8Br2 Formula Weight: 215.93 Flash Point: None Refractive Index: 1.5095 Boiling Point: 173-176° Beilstein Reference: 1731231 MDL No.: MFCD00000152

Formula: BrH₂CCH₂BrCHCH₃
MW: 215.92 g/mol
Boiling Pt: 173…176 °C
Density: 1.789
Storage Temperature: Ambient
MDL Number: MFCD00000152
CAS Number: 107-80-2
EINECS: 203-520-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR