2,5-Difluorophenylboronic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24113-03
B24113-06
B24113-14
CAAAB24113-03EA
43.13
CAD
CAAAB24113-03
CAAAB24113-06
CAAAB24113-14
2,5-Difluorophenylboronic acid 96%
2,5-Difluorophenylboronic acid
Purity: 96%
CAS No.: 193353-34-3
Molecular Formula: C6H5BF2O2
Formula Weight: 157.91
Beilstein Reference: 8833254
MDL No.: MFCD01863171
Formula:
C₆H₅BF₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01863171 CAS Number: 193353-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...