1-Amino-3-methoxypropane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methoxypropylamine

B24095-AK B24095-AU
CAAAB24095-AKEA 48.98 CAD
CAAAB24095-AK AAB24095-AU
1-Amino-3-methoxypropane 99%
1-Amino-3-methoxypropane

Purity: 99% CAS No.: 5332-73-0 Molecular Formula: CH3O(CH2)3NH2 Formula Weight: 89.14 Flash Point: 27°(81°F) Refractive Index: 1.4175 Odor: Amine-like Melting Point: -70° Boiling Point: 117-119° Beilstein Reference: 878144 UN No.: 2734 MDL No.: MFCD00014831

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 117…119 °C
Melting Pt: < –70 °C
Density: 0.872
Flash Pt: 27 °C(81 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014831
CAS Number: 5332-73-0
EINECS: 226-241-3
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR