4-Chlororesorcinol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chlorobenzene-1,3-diol, 1-Chloro-2,4-dihydroxybenzene

B24076-14 B24076-22 B24076-36
CAAAB24076-14EA 45.03 CAD
CAAAB24076-14 CAAAB24076-22 CAAAB24076-36
4-Chlororesorcinol 98%
4-Chlororesorcinol

Purity: 98% CAS No.: 95-88-5 Molecular Formula: C6H5ClO2 Formula Weight: 144.56 Melting Point: 108-111° Boiling Point: 146-148°/18mm MDL No.: MFCD00002273

Formula: C₆H₅ClO₂
MW: 144.56 g/mol
Boiling Pt: 146…148 °C/18 mm
Melting Pt: 108…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00002273
CAS Number: 95-88-5
EINECS: 202-462-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR