1H,1H,2H,2H-Perfluoro-1-iodohexane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24075-06
B24075-14
CAAAB24075-06EA
104.75
CAD
CAAAB24075-06
CAAAB24075-14
1H,1H,2H,2H-Perfluoro-1-iodohexane 97%
1H,1H,2H,2H-Perfluoro-1-iodohexane
Purity: 97%
CAS No.: 2043-55-2
Molecular Formula: CF3(CF2)3CH2CH2I
Formula Weight: 373.99
Flash Point:
Formula:
C₆H₄F₉I Boiling Pt: 138…140 °C Melting Pt: –25 °C Density: 1.940 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039409 CAS Number: 2043-55-2 EINECS: 218-055-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...