1,3-Benzenedisulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzene-1,3-di(sulfonyl chloride), Benzene-1,3-disulfonyl chloride, Benzene-1,3-di(sulphonyl chloride), Benzene-1,3-disulphonyl chloride

B24044-06 B24044-14 B24044-22
CAAAB24044-06EA 117.24 CAD
CAAAB24044-06 CAAAB24044-14 CAAAB24044-22
1,3-Benzenedisulfonyl chloride 98%
1,3-Benzenedisulphonyl chloride

Purity: 98% CAS No.: 585-47-7 Molecular Formula: C6H4Cl2O4S2 Formula Weight: 275.13 Melting Point: 58-60° Beilstein Reference: 1118508 UN No.: 3261 MDL No.: MFCD00041879

Formula: C₆H₄Cl₂O₄S₂
MW: 275.13 g/mol
Boiling Pt: 145 °C (1 -rr)
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00041879
CAS Number: 585-47-7
EINECS: 209-556-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR