Ethylurea 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Ethylurea

B24040-22 B24040-36
CAAAB24040-22EA 271.76 CAD
CAAAB24040-22 CAAAB24040-36
Ethylurea 98%
Ethylurea

Purity: 98% CAS No.: 625-52-5 Molecular Formula: CH3CH2NHCONH2 Formula Weight: 88.11 Melting Point: 88-94° Beilstein Reference: 1740512

Formula: C₃H₈N₂O
MW: 88.11 g/mol
Boiling Pt: 258
Melting Pt: 94.55
Storage Temperature: Ambient
MDL Number: MFCD00007953
CAS Number: 625-52-5
EINECS: 210-898-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR