N,N-Dimethylethylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

B23992-AE B23992-AP
CAAAB23992-AEEA 73.47 CAD
CAAAB23992-AE CAAAB23992-AP
N,N-Dimethylethylamine 99%
N,N-Dimethylethylamine

Purity: 99% CAS No.: 598-56-1 Molecular Formula: C4H11N Formula Weight: 73.14 Storage Temperature: 2°C to 8°C Flash Point: -36°(-33°F) Refractive Index: 1.3730 Odor: Ammonia-like Melting Point: -140° Boiling Point: 36-37° Beilstein Reference: 1696893 UN No.: 2733 MDL No.: MFCD00009039

Formula: C₄H₁₁N
Boiling Pt: 36…37 °C
Melting Pt: –140 °C
Density: 0.675
Flash Pt: –36 °C (–33 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009039
CAS Number: 598-56-1
EINECS: 209-940-8
UN: 2733
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR