2-Bromophenylhydrazinium chloride 95%

Supplier: Thermo Scientific Chemicals
Warning

B23966-06 B23966-14 B23966-22
CAAAB23966-06EA 114.23 CAD
CAAAB23966-06 CAAAB23966-14 CAAAB23966-22
2-Bromophenylhydrazinium chloride 95%
2-Bromophenylhydrazinium chloride

Purity: 95% CAS No.: 50709-33-6 Molecular Formula: C6H7BrN2.HCl Formula Weight: 223.51 Melting Point: ca 188° dec. MDL No.: MFCD00012926

Formula: C₆H₈BrClN₂
Melting Pt: ca 188 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012926
CAS Number: 50709-33-6
EINECS: 256-728-6
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR