3-Toluoyl chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23914-14
B23914-22
B23914-36
CAAAB23914-14EA
49.45
CAD
CAAAB23914-14
CAAAB23914-22
CAAAB23914-36
3-Toluoyl chloride 99%
3-Toluoyl chloride
Purity: 99%
CAS No.: 1711-06-4
Molecular Formula: C8H7ClO
Formula Weight: 154.60
Flash Point: 76°(168°F)
Refractive Index: 1.5485
Odor: Pungent
Melting Point: -23°
Boiling Point: 220°
Beilstein Reference: 878419
UN No.: 3265
MDL No.: MFCD00000681
Formula:
C₈H₇ClO Boiling Pt: 220 °C Melting Pt: –23 °C Density: 1.173 Flash Pt: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000681 CAS Number: 1711-06-4 EINECS: 216-976-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...