Symclosene 90+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichloro-1,3,5-triazinetrion, 1,3,5-Trichlorohexahydro-1,3,5-triazine-2,4,6-trione, Trichlorocyanuric acid, TCCA, 1,3,5-Trichloro-1,3,5-triazine-2,4,6-trione, Trichloroisocyanuric acid

B23906-0E B23906-22 B23906-36
CAAAB23906-0EEA 364.98 CAD
CAAAB23906-0E CAAAB23906-22 CAAAB23906-36
Symclosene 90+%
Symclosene

Purity: 90+% CAS No.: 87-90-1 Molecular Formula: C3Cl3N3O3 Formula Weight: 232.41 Flash Point: 250°(482°F) Odor: Pungent pH: 3 Melting Point: ca 245° dec. Beilstein Reference: 202022 Merck Reference: 14,9641 UN No.: 2468 MDL No.: MFCD00006553

Formula: C₃Cl₃N₃O₃
MW: 232.41 g/mol
Boiling Pt: 413
Melting Pt: ∼ 245 °C (dec.)
Density: 2.07
Flash Pt: >250 °C (482 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006553
CAS Number: 87-90-1
EINECS: 201-782-8
UN: 2468
ADR: 5.1 II
Merck Index: 14,9641

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR