1,8-Diaminooctane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23885-14
B23885-22
CAAAAB23885-14EA
76.31
CAD
CAAAAB23885-14
CAAAAB23885-22
1,8-Diaminooctane 98%
1,8-Diaminooctane
Purity: 98%
CAS No.: 373-44-4
Molecular Formula: H2N(CH2)8NH2
Formula Weight: 144.26
Flash Point: 106°(223°F)
Odor: Amine-like
pH: 11.7
Melting Point: 50-54°
Boiling Point: 225-226°
Beilstein Reference: 1735426
UN No.: 3259
MDL No.: MFCD00008248
Formula:
C₈H₂₀N₂ Boiling Pt: 225…226 °C Melting Pt: 50…54 °C Density: 0.83 g/cm³ (60 °C) Flash Pt: 106 °C (223 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008248 CAS Number: 373-44-4 EINECS: 206-764-3 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...