(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(-)-BINAP, (S)-BINAP, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (S)-(-)-2,2'-Bis(diphenylphoshino)-1,1'-binaphthyl
B23872-03
B23872-06
CAAAAB23872-01EA
71.98
CAD
CAAAAB23872-01
CAAAAB23872-03
CAAAAB23872-06
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Purity: 97%
CAS No.: 76189-56-5
Molecular Formula: C44H32P2
Formula Weight: 622.69
Melting Point: 239-241°
Beilstein Reference: 5321444
Merck Reference: 14,1223
Optical Rotation: -230° (c=0.3 in toluene)
MDL No.: MFCD00010805
Formula:
C₄₄H₃₂P₂ MW: 622.69 g/mol Melting Pt: 239…241 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010805 CAS Number: 76189-56-5 Merck Index: 14,1223 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...