4-Chlorothiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol
B23838-14
B23838-22
CAAAB23838-14EA
47.56
CAD
CAAAB23838-14
CAAAB23838-22
CAAAB23838-36
4-Chlorothiophenol 97%
4-Chlorothiophenol
Purity: 97%
CAS No.: 106-54-7
Molecular Formula: C6H5ClS
Formula Weight: 144.62
Flash Point: 81°(177°F)
Melting Point: 48-52°
Boiling Point: 205-207°
Beilstein Reference: 605971
UN No.: 1759
MDL No.: MFCD00004847
Formula:
C₆H₅ClS Boiling Pt: 205…207 °C Melting Pt: 48…52 °C Density: 1.224 g/cm³ (60 °C) Flash Pt: 81 °C (177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004847 CAS Number: 106-54-7 EINECS: 203-408-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...