3-Cyanobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23836-06
B23836-14
B23836-22
CAAAB23836-06EA
82.16
CAD
CAAAB23836-06
CAAAB23836-14
CAAAB23836-22
3-Cyanobenzoic acid 98+%
3-Cyanobenzoic acid
Purity: 98+%
CAS No.: 1877-72-1
Molecular Formula: C8H5NO2
Formula Weight: 147.13
Melting Point: 219-224°
Beilstein Reference: 1862566
Formula:
C₈H₅NO₂ Melting Pt: 219…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002486 CAS Number: 1877-72-1 EINECS: 217-511-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...