Diethyl diethylmalonate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Diethyl Diethylmalonate
B23810-14
B23810-22
B23810-36
CAAAB23810-14EA
73.15
CAD
CAAAB23810-14
CAAAB23810-22
CAAAB23810-36
Diethyl diethylmalonate 98%
Diethyl diethylmalonate
Purity: 98%
CAS No.: 77-25-8
Molecular Formula: C11H20O4
Formula Weight: 216.28
Flash Point: 94°(202°F)
Refractive Index: 1.4230
Boiling Point: 228-230°
Merck Reference: 14,3789
MDL No.: MFCD00009132
Formula:
C₁₁H₂₀O₄ Boiling Pt: 228…230 °C Density: 0.990 Flash Pt: 94 °C(202 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009132 CAS Number: 77-25-8 EINECS: 201-016-2 Merck Index: 14,3789 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...