1,8-Nonadiyne 97%

Supplier: Thermo Scientific Chemicals
Warning

B23784-06 B23784-14
CAAAB23784-06EA 131.14 CAD
CAAAB23784-06 CAAAB23784-14
1,8-Nonadiyne 97%
1,8-Nonadiyne

Purity: 97% CAS No.: 2396-65-8 Molecular Formula: C9H12 Formula Weight: 120.20 Flash Point: 41°(105°F) Refractive Index: 1.4490 Odor: Characteristic Melting Point: -21° Boiling Point: 55-56°/13mm Beilstein Reference: 1733443 UN No.: 3295 MDL No.: MFCD00008581

Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 55…56 °C/13 mm
Melting Pt: –21 °C
Density: 0.799
Flash Pt: 41 °C(105 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008581
CAS Number: 2396-65-8
EINECS: 219-254-0
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR