Caproic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23756-0B
B23756-18
B23756-30
CAAAB23756-0BEA
355.5
CAD
CAAAB23756-0B
CAAAB23756-18
CAAAB23756-30
Caproic anhydride 98%
Caproic anhydride
Purity: 98%
CAS No.: 2051-49-2
Molecular Formula: C12H22O3
Formula Weight: 214.31
Flash Point: 128°(262°F)
Refractive Index: 1.4290
Odor: Unpleasant
Melting Point: -40°
Boiling Point: 246-248°
Beilstein Reference: 1776561
UN No.: 3265
MDL No.: MFCD00009509
Formula:
C₁₂H₂₂O₃ Boiling Pt: 246…248 °C Melting Pt: –40 °C Density: 0.928 Flash Pt: 128 °C (262 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009509 CAS Number: 2051-49-2 EINECS: 218-121-4 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...