Octyltrichlorosilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23753-14
B23753-22
B23753-36
CAAAB23753-14EA
52.46
CAD
CAAAB23753-14
CAAAB23753-22
CAAAB23753-36
Octyltrichlorosilane 97%
Octyltrichlorosilane
Purity: 97%
CAS No.: 5283-66-9
Molecular Formula: CH3(CH2)7SiCl3
Formula Weight: 247.67
Flash Point: 96°(204°F)
Refractive Index: 1.4470
Odor: Irritating
Boiling Point: 225-227°
Beilstein Reference: 1744230
UN No.: 1801
MDL No.: MFCD00000488
Formula:
C₈H₁₇Cl₃Si Boiling Pt: 225…227 °C Density: 1.073 Flash Pt: 96 °C (204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000488 CAS Number: 5283-66-9 EINECS: 226-112-1 UN: 1801 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...