(3-Mercaptopropyl)trimethoxysilane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23726-14
B23726-22
B23726-36
CAAAAB23726-14EA
59.57
CAD
CAAAAB23726-14
CAAAAB23726-22
CAAAAB23726-36
(3-Mercaptopropyl)trimethoxysilane 95%
(3-Mercaptopropyl)trimethoxysilane
Purity: 95%
CAS No.: 4420-74-0
Molecular Formula: C6H16O3SSi
Formula Weight: 196.34
Flash Point: 96°(204°F)
Refractive Index: 1.4430
Melting Point:
Formula:
HSH₂CCH₂CH₂Si(OCH₃)₃ Boiling Pt: 213…215 °C Melting Pt: < –50 °C Density: 1.050 Flash Pt: 96 °C(204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004901 CAS Number: 4420-74-0 EINECS: 224-588-5 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...