Ethanesulfonyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23723-14
B23723-22
B23723-36
CAAAB23723-14EA
50.4
CAD
CAAAB23723-14
CAAAB23723-22
CAAAB23723-36
Ethanesulfonyl chloride 98+%
Ethanesulphonyl chloride
Purity: 98+%
CAS No.: 594-44-5
Molecular Formula: C2H5ClO2S
Formula Weight: 128.57
Flash Point: 83°(181°F)
Refractive Index: 1.4540
Melting Point: -70°
Boiling Point: 176-178°
Beilstein Reference: 773865
UN No.: 2927
MDL No.: MFCD00007460
Formula:
C₂H₅ClO₂S Boiling Pt: 176…178 °C Melting Pt: –70 °C Density: 1.350 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007460 CAS Number: 594-44-5 EINECS: 209-842-5 UN: 2927 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...