Manganese(II) acetate, anhydrous 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid manganese (II) salt, Manganese diacetate

B23708-0B B23708-18 B23708-30
CAAAAB23708-0BEA 948 CAD
CAAAAB23708-0B CAAAAB23708-18 CAAAAB23708-30
Manganese(II) acetate, anhydrous 98+%
Manganese (II) acetate

Purity: 98+% CAS No.: 638-38-0 Molecular Formula: C4H6MnO4 Formula Weight: 173.03 Merck Reference: 14,5724 MDL No.: MFCD00013039

Formula: (H₃CCOO)₂Mn
MW: 173.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013039
CAS Number: 638-38-0
EINECS: 211-334-3
Merck Index: 14,5724

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR