1,1,3,3-Tetramethyldisiloxane 97%

Supplier: Thermo Scientific Chemicals
Danger

B23697-36 B23697-14 B23697-22
CAAAAB23697-36EA 820.02 CAD
CAAAAB23697-36 CAAAB23697-14 CAAAB23697-22
1,1,3,3-Tetramethyldisiloxane 97%
1,1,3,3-Tetramethyldisiloxane

Purity: 97% CAS No.: 3277-26-7 Molecular Formula: C4H14OSi2 Formula Weight: 134.33 Flash Point: -26°(-15°F) Refractive Index: 1.3700 Odor: Characteristic Melting Point: -78° Boiling Point: 70-71° Beilstein Reference: 1733364 UN No.: 1993 MDL No.: MFCD00008256

Formula: C₄H₁₄OSi₂
Boiling Pt: 70…71 °C
Melting Pt: –78 °C
Density: 0.760
Flash Pt: –26 °C (–15 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008256
CAS Number: 3277-26-7
EINECS: 221-906-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR