2-Methylfuran 98+% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23692-AK
B23692-AU
CAAAB23692-AKEA
112.97
CAD
CAAAB23692-AK
CAAAB23692-AU
2-Methylfuran 98+% stabilized
2-Methylfuran
Purity: 98+%
CAS No.: 534-22-5
Molecular Formula: C5H6O
Formula Weight: 82.10
Flash Point: -30°(-22°F)
Refractive Index: 1.4330
Melting Point: -89°
Boiling Point: 63-65°
Beilstein Reference: 103733
UN No.: 2301
MDL No.: MFCD00003248
Formula:
C₅H₆O MW: 82.1 g/mol Boiling Pt: 63…65 °C Melting Pt: –89 °C Density: 0.910 Flash Pt: –30 °C (–22 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003248 CAS Number: 534-22-5 EINECS: 208-594-5 UN: 2301 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...