2-(Dimethylamino)-ethanol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Dimethylaminoethanol, N,N-Dimethylethanolamine, DMAE

B23616-AK B23616-AU
CAAAB23616-AKEA 38.55 CAD
CAAAB23616-AK CAAAB23616-AU
2-(Dimethylamino)-ethanol 99+%
2-Dimethylaminoethanol

Purity: 99+% CAS No.: 108-01-0 Molecular Formula: C4H11NO Formula Weight: 89.14 Flash Point: 40°(104°F) Refractive Index: 1.4295 Odor: Ammonia-like Melting Point: -60° Boiling Point: 134-136° Beilstein Reference: 1209235 Merck Reference: 14,2843 UN No.: 2051 MDL No.: MFCD00002846

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 134…136 °C
Melting Pt: -59
Density: 0.886
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002846
CAS Number: 108-01-0
EINECS: 203-542-8
UN: 2051
ADR: 8 II
Merck Index: 14,2843

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR