Tin(II) 2-ethylhexanoate 95% (Sn basis)

Supplier: Thermo Scientific Chemicals
Danger

B23612-14 B23612-22
CAAAAB23612-14EA 71.26 CAD
CAAAAB23612-14 CAAAAB23612-22
Tin(II) 2-ethylhexanoate 95% (Sn basis)
Tin (II) 2-ethylhexanoate

Purity: 96% CAS No.: 301-10-0 Molecular Formula: C16H30O4Sn Formula Weight: 405.11 Flash Point: 110°(230°F) Refractive Index: 1.4940 Odor: Characteristic MDL No.: MFCD00002676

Formula: C₁₆H₃₀O₄Sn
Density: 1.251
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6

Order Now

Specification Test Results

Appearance (Color) Clear colorless to yellow
Form Viscous liquid
Assay from Supplier's CofA ≥94.0 to ≤106.0% (based on Sn EA)
Elemental Analysis: Carbon (C) 44.59-50.28%
Elemental Analysis: Tin (Sn) 27.55-31.06%
Identification (FTIR) Conforms
Refractive Index 1.4910-1.4970 @ 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR