2-Chlorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23596-22
B23596-0B
CAAAAB23596-22EA
43.92
CAD
CAAAAB23596-22
CAAAB23596-0B
2-Chlorotoluene 98%
2-Chlorotoluene
Purity: 98%
CAS No.: 95-49-8
Molecular Formula: C7H7Cl
Formula Weight: 126.59
Flash Point: 47°(116°F)
Refractive Index: 1.5250
Melting Point: -36°
Boiling Point: 157-159°
Beilstein Reference: 1904175
Merck Reference: 14,2171
UN No.: 2238
MDL No.: MFCD00000562
Formula:
C₇H₇Cl Boiling Pt: 157…159 °C Melting Pt: –36 °C Density: 1.082 Flash Pt: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000562 CAS Number: 95-49-8 EINECS: 202-424-3 UN: 2238 ADR: 3,III Merck Index: 14,2171 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...