N-Ethylethylenediamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Aminoethyl(ethyl)amine
B23577-06
B23577-14
CAAAB23577-06EA
42.5
CAD
CAAAB23577-06
CAAAB23577-14
N-Ethylethylenediamine 98%
N-Ethylethylenediamine
Purity: 98%
CAS No.: 110-72-5
Molecular Formula: CH3CH2NHCH2CH2NH2
Formula Weight: 88.15
Flash Point: 34°(93°F)
Refractive Index: 1.4400
Boiling Point: 128-130°
Beilstein Reference: 1098272
UN No.: 2733
MDL No.: MFCD00008166
Formula:
C₄H₁₂N₂ Boiling Pt: 128…130 °C Density: 0.837 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008166 CAS Number: 110-72-5 EINECS: 203-795-4 UN: 2733 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...