3-Anisoyl chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23571-09
B23571-18
B23571-30
CAAAB23571-09EA
85.79
CAD
CAAAB23571-09
CAAAB23571-18
CAAAB23571-30
3-Anisoyl chloride 99%
3-Anisoyl chloride
Purity: 99%
CAS No.: 1711-05-3
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Flash Point: 92°(197°F)
Refractive Index: 1.5586
Boiling Point: 123-125°/15mm
Beilstein Reference: 386659
UN No.: 1729
MDL No.: MFCD00000673
Formula:
C₈H₇ClO₂ Boiling Pt: 123…125 °C/15 mm Density: 1.214 Flash Pt: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000673 CAS Number: 1711-05-3 EINECS: 216-975-2 UN: 1729 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...