Chlorobutyldimethylsilane 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: B 2785, Butylchlorodimethylsilane, n-Butyldimethylchlorosilane, Silan BM2, Butyldimethylchlorosilane

B23551-06 B23551-14 B23551-22
CAAAAB23551-06EA 54.98 CAD
CAAAAB23551-06 CAAAAB23551-14 CAAAAB23551-22
Chlorobutyldimethylsilane 96%
Chlorobutyldimethylsilane

Purity: 96% CAS No.: 1000-50-6 Molecular Formula: CH3(CH2)3Si(CH3)2Cl Formula Weight: 150.73 Flash Point: 27°(80°F) Refractive Index: 1.4200 Boiling Point: 138° Beilstein Reference: 1732573 UN No.: 2920 MDL No.: MFCD00015727

Formula: C₆H₁₅ClSi
MW: 150.72 g/mol
Boiling Pt: 138 °C
Density: 0.875
Flash Pt: 27 °C (80 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015727
CAS Number: 1000-50-6
EINECS: 423-700-1
UN: 2986
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR