1,2-Dimethylimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Dimethyl-1H-imidazole

B23544-14 B23544-22 B23544-36
CAAAAB23544-14EA 35.87 CAD
CAAAAB23544-14 CAAAAB23544-22 CAAAAB23544-36
1,2-Dimethylimidazole 98%
1,2-Dimethylimidazole

Purity: 98% CAS No.: 1739-84-0 Molecular Formula: C5H8N2 Formula Weight: 96.13 Flash Point: 92°(197°F) Melting Point: 35-38° Boiling Point: 203-205° Beilstein Reference: 108457 MDL No.: MFCD00005294

Formula: C₅H₈N₂
Boiling Pt: 203…205 °C
Melting Pt: 35…38 °C
Density: 1,084
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005294
CAS Number: 1739-84-0
EINECS: 217-101-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR