3-Bromofuran 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with calcium carbonate 0.5%

B23543-03 B23543-06 B23543-14
CAAAAB23543-03EA 94.8 CAD
CAAAAB23543-03 CAAAAB23543-06 CAAAAB23543-14
3-Bromofuran 97% stabilized
3-Bromofuran

Purity: 97% CAS No.: 22037-28-1 Molecular Formula: C4H3BrO Formula Weight: 146.98 Storage Temperature: -30°C to -10°C Flash Point: 3°(37°F) Refractive Index: 1.4960 Boiling Point: 102-104° Beilstein Reference: 105337 UN No.: 1993 MDL No.: MFCD00005347

Formula: C₄H₃BrO
Boiling Pt: 102…104 °C
Density: 1.635
Flash Pt: 3 °C (37 °F)
Storage Temperature: Freezer
MDL Number: MFCD00005347
CAS Number: 22037-28-1
EINECS: 244-744-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR